Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004262.2 | NP_004253.1 | 3 | 294 | ||
XM_005265710.4 | XP_005265767.1 | 2 | 1237 | |||
GenBank mRNA | AB002134.1 | - | 3 | 290 | ||
AK290289.1 | - | 3 | 295 | |||
AK296853.1 | - | 2 | 157 | |||
AL603214.1 | - | 3 | 294 | |||
BC125195.1 | - | 3 | 279 | 40123006 | ||
BC125196.1 | - | 3 | 279 | 40123011 |