Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000221.2 | NP_000212.1 | 5, 6 | 1086 | ||
NM_006488.2 | NP_006479.1 | 5, 6 | 1086 | |||
XM_005264294.3 | XP_005264351.1 | 6, 7 | 1229 | |||
XM_005264296.3 | XP_005264353.1 | 5, 6 | 1072 | |||
XM_005264298.3 | XP_005264355.1 | 3, 4 | 839 | |||
XM_017004060.1 | XP_016859549.1 | 6, 7 | 1226 | |||
XM_017004061.1 | XP_016859550.1 | 4, 5 | 956 | |||
GenBank mRNA | AK130033.1 | - | 5, 6 | 603 | ||
AK223075.1 | - | 5, 6 | 603 | |||
BC006233.2 | - | 5, 6 | 1024 | 3946893 | ||
BX648873.1 | - | 3, 4 | 353 | |||
CR456801.1 | - | 5, 6 | 569 | |||
X78677.1 | - | 3, 4 | 365 | |||
X78678.1 | - | 5, 6 | 577 |