Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 4 | 1147 | ||
NM_001167607.1 | NP_001161079.1 | 4 | 937 | |||
GenBank mRNA | AK225978.1 | - | 4 | 1075 | ||
BC009273.2 | - | 4 | 1010 | 4053681 | ||
X61095.1 | - | 4 | 1004 | |||
X62078.1 | - | 4 | 1040 |