Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 4 | 2511 | ||
NM_001167607.1 | NP_001161079.1 | 4 | 2301 | |||
GenBank mRNA | BC009273.2 | - | 4 | 2372 | 4053681 | |
X62078.1 | - | 4 | 2405 |