Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000572.2 | NP_000563.1 | 5 | 974 | ||
GenBank mRNA | M57627.1 | - | 5 | 945 |