Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000583.3 | NP_000574.2 | 4 | 745 | ||
NM_001204306.1 | NP_001191235.1 | 5 | 616 | |||
NM_001204307.1 | NP_001191236.1 | 5 | 553 | |||
XM_006714177.2 | XP_006714240.1 | 4 | 608 | |||
GenBank mRNA | AK223458.1 | - | 5 | 616 | ||
AK290827.1 | - | 4 | 455 | |||
AK298433.1 | - | 3 | 313 | |||
AK309595.1 | - | 5 | 553 | |||
AK312971.1 | - | 4 | 459 | |||
AK315853.1 | - | 4 | 608 | |||
BC057228.1 | - | 4 | 463 | 4767205 | ||
EU794674.1 | - | 4 | 554 | |||
X03178.1 | - | 4 | 554 |