Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000586.3 | NP_000577.2 | 3 | 324 | ||
XM_017008177.1 | XP_016863666.1 | 3 | 290 | |||
GenBank mRNA | A14844.1 | - | 3 | 279 | ||
AB451325.1 | - | 3 | 269 | |||
AB451454.1 | - | 3 | 269 | |||
AF228636.1 | - | 3 | 281 | |||
AY523040.1 | - | 3 | 209 | |||
BC066254.1 | - | 3 | 287 | 6971690 | ||
BC066255.1 | - | 3 | 287 | 6971691 | ||
BC066256.1 | - | 3 | 287 | 6971693 | ||
BC066257.1 | - | 3 | 287 | 6971694 | ||
BC070338.1 | - | 3 | 316 | 30418817 | ||
DQ231169.1 | - | 3 | 209 | |||
DQ861285.1 | - | 3 | 271 | |||
S77834.1 | - | 3 | 316 | |||
S77835.1 | - | 3 | 258 | |||
S82692.1 | - | 3 | 563 | |||
V00564.1 | - | 3 | 316 | |||
X01586.1 | - | 3 | 376 |