Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000590.1 | NP_000581.1 | 4 | 213 | ||
GenBank mRNA | BC066283.1 | - | 4 | 213 | 6971717 | |
BC066284.1 | - | 4 | 213 | 6971718 | ||
BC066285.1 | - | 4 | 213 | 6971719 | ||
BC066286.1 | - | 4 | 213 | 6971720 | ||
BC066287.1 | - | 4 | 213 | 6971721 | ||
M30134.1 | - | 4 | 213 | |||
S63356.1 | - | 4 | 213 | |||
X17543.1 | - | 4 | 213 |