Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000590.1 | NP_000581.1 | 5 | 533 | ||
GenBank mRNA | M30134.1 | - | 5 | 533 | ||
S63356.1 | - | 5 | 533 | |||
X17543.1 | - | 5 | 533 |