Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000824.4 | NP_000815.1 | 10 | 1864 | ||
NM_001166060.1 | NP_001159532.1 | 10 | 1847 | |||
NM_001166061.1 | NP_001159533.1 | 9 | 1571 | |||
XM_017008034.1 | XP_016863523.1 | 9 | 1658 | |||
XM_017008035.1 | XP_016863524.1 | 9 | 1548 | |||
GenBank mRNA | AK290617.1 | - | 10 | 1798 | ||
BC032635.1 | - | 10 | 1803 | 5247588 | ||
U33267.1 | - | 10 | 1738 |