Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000839.3 | NP_000830.2 | 4 | 1846 | ||
NM_001130063.1 | NP_001123535.1 | 2 | 422 | |||
XM_011533635.1 | XP_011531937.1 | 3 | 3421 | |||
XM_011533636.1 | XP_011531938.1 | 3 | 2971 | |||
XM_011533637.1 | XP_011531939.1 | 3 | 1842 | |||
XM_011533638.2 | XP_011531940.1 | 4 | 1584 | |||
XM_011533639.2 | XP_011531941.1 | 4 | 1302 | |||
XM_011533640.2 | XP_011531942.1 | 4 | 1255 | |||
XM_011533641.2 | XP_011531943.1 | 4 | 1291 | |||
XM_011533643.1 | XP_011531945.1 | 4 | 460 | |||
XM_011533644.1 | XP_011531946.1 | 3 | 536 | |||
XM_011533645.1 | XP_011531947.1 | 3 | 509 | |||
XM_011533646.2 | XP_011531948.1 | 3 | 712 | |||
XM_011533647.1 | XP_011531949.1 | 4 | 600 | |||
XM_017006271.1 | XP_016861760.1 | 4 | 2324 | |||
GenBank mRNA | AB209164.1 | - | 3 | 4087 | ||
AK289984.1 | - | 4 | 1839 | |||
AK313056.1 | - | 5 | 1981 | |||
AK315816.1 | - | 4 | 1841 | |||
AY999299.1 | - | 3 | 1643 | |||
BC041447.1 | - | 2 | 397 | |||
BC113615.1 | - | 3 | 1630 | 8322667 | ||
BC113619.1 | - | 3 | 1630 | 8322671 | ||
BX648951.1 | - | 2 | 5037 | |||
EU432122.1 | - | 3 | 1607 | |||
L35318.1 | - | 3 | 1609 |
Set Membership: |
G-protein coupled receptor |