Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000841.3 | NP_000832.1 | 11 | 3867 | ||
NM_001256809.2 | NP_001243738.1 | 9 | 3250 | |||
NM_001256811.2 | NP_001243740.1 | 10 | 3726 | |||
NM_001256812.2 | NP_001243741.1 | 10 | 2951 | |||
NM_001256813.2 | NP_001243742.1 | 11 | 3175 | |||
NM_001282847.1 | NP_001269776.1 | 10 | 3650 | |||
XM_017010790.1 | XP_016866279.1 | 11 | 3805 | |||
XM_017010791.1 | XP_016866280.1 | 11 | 3860 | |||
XM_017010792.1 | XP_016866281.1 | 10 | 3404 | |||
XM_017010793.1 | XP_016866282.1 | 10 | 3155 | |||
GenBank mRNA | AK122982.1 | - | 11 | 3160 | ||
AK126746.1 | - | 10 | 3415 | |||
AK131536.1 | - | 9 | 3250 | |||
AK289524.1 | - | 4185 | ||||
U92457.1 | - | 10 | 3390 | |||
X80818.1 | - | 10 | 3536 |
Set Membership: |
G-protein coupled receptor |