Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000860.5 | NP_000851.2 | 6, 7 | 1118 | ||
NM_001256301.1 | NP_001243230.1 | 6, 7 | 848 | |||
NM_001256306.1 | NP_001243235.1 | 4, 5 | 914 | |||
NM_001256307.1 | NP_001243236.1 | 5, 6 | 1011 | |||
GenBank mRNA | AK058013.1 | - | 6, 7 | 707 | ||
AK296642.1 | - | 6, 7 | 848 | |||
AK300125.1 | - | 4, 5 | 504 | |||
AK300524.1 | - | 5, 6 | 754 | |||
AK314624.1 | - | 6, 7 | 707 | |||
BC018986.2 | - | 6, 7 | 680 | 3638799 | ||
J05594.1 | - | 6, 7 | 688 | |||
L76465.1 | - | 6, 7 | 688 |