Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000866.4 | NP_000857.1 | 2 | 512 | ||
NM_001322208.1 | NP_001309137.1 | 3 | 793 | |||
NM_001322209.1 | NP_001309138.1 | 3 | 589 | |||
NM_001322210.1 | NP_001309139.1 | 3 | 580 | |||
XM_005264751.3 | XP_005264808.1 | 3 | 659 | |||
XM_011533664.2 | XP_011531966.1 | 3 | 591 | |||
GenBank mRNA | AF498981.1 | - | 1 | 354 | ||
BC069125.1 | - | 1 | 354 | 7216980 | ||
BX357988.2 | - | 2 | 512 |
Set Membership: |
G-protein coupled receptor |