Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135650.1 | NP_001129122.1 | 2 | 315 | ||
NM_004280.4 | NP_004271.1 | 2 | 315 | |||
GenBank mRNA | AB011079.1 | - | 2 | 241 | ||
AF054186.1 | - | 2 | 269 | |||
AV714140.2 | - | 2 | 315 | |||
BC005291.1 | - | 2 | 268 | 3685030 | ||
BP235196.1 | - | 2 | 298 | |||
BT007306.1 | - | 2 | 241 | |||
CK002875.1 | - | 2 | 269 |
Gene Symbol: EEF1E1-BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037618.1 | 2 | 315 | |||
GenBank mRNA | BI552521.1 | - | 2 | 255 |