Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000888.4 | NP_000879.2 | 4 | 758 | ||
NM_001282353.1 | NP_001269282.1 | 5 | 651 | |||
NM_001282354.1 | NP_001269283.1 | 2 | 473 | |||
NM_001282355.1 | NP_001269284.1 | 4 | 758 | |||
NM_001282388.1 | NP_001269317.1 | 3 | 678 | |||
NM_001282389.1 | NP_001269318.1 | 4 | 585 | |||
NM_001282390.1 | NP_001269319.1 | 3 | 505 | |||
GenBank mRNA | AK290300.1 | - | 4 | 726 | ||
AK304203.1 | - | 3 | 617 | |||
AK313944.1 | - | 5 | 642 | |||
AL831998.1 | - | 4 | 697 | |||
BC121178.2 | - | 4 | 528 | 40122759 | ||
BC143656.1 | - | 4 | 536 | |||
CD013944.1 | - | 3 | 450 | |||
CD013945.1 | - | 4 | 703 | |||
CD013946.1 | - | 2 | 418 | |||
CD013947.1 | - | 4 | 530 | |||
M35198.3 | - | 4 | 524 |