Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004286.4 | NP_004277.2 | 6, 7 | 1235 | ||
XM_011530537.2 | XP_011528839.1 | 6, 7 | 2497 | |||
XM_017029099.1 | XP_016884588.1 | 6, 7 | 2489 | |||
XM_017029100.1 | XP_016884589.1 | 6, 7 | 2459 | |||
XM_017029101.1 | XP_016884590.1 | 7, 8 | 1076 | |||
XM_017029102.1 | XP_016884591.1 | 5, 6 | 892 | |||
XM_017029103.1 | XP_016884592.1 | 5, 6 | 990 | |||
GenBank mRNA | AK315704.1 | - | 5, 6 | 858 | ||
BC014075.1 | - | 6, 7 | 1073 | |||
CR456501.1 | - | 6, 7 | 1058 | |||
U87964.1 | - | 6, 7 | 979 |