Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF14 SIRNA Assays ›
Gene Symbol:
RNF14
Gene Name:
ring finger protein 14
Gene Aliases:
ARA54, HFB30, HRIHFB2038, TRIAD2
Chromosome Location:
Chr.5: 141949337 - 141993709 on Build GRCh38
UniGene ID:
Hs.483616
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14469
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201365.1
NP_001188294.1
3
366
NM_004290.4
NP_004281.1
3
580
NM_183398.2
NP_899645.1
3
580
NM_183399.2
NP_899646.1
2
192
NM_183400.2
NP_899647.1
2
406
NM_183401.2
NP_899648.1
3
386
XM_005268536.2
XP_005268593.1
3
683
XM_005268537.2
XP_005268594.1
3
727
XM_005268539.2
XP_005268596.1
3
348
XM_005268540.3
XP_005268597.1
3
504
XM_005268541.3
XP_005268598.1
2
260
XM_005268542.2
XP_005268599.1
3
423
XM_005268543.4
XP_005268600.1
2
229
XM_011537714.2
XP_011536016.1
2
197
XM_017010070.1
XP_016865559.1
3
331
XM_017010071.1
XP_016865560.1
3
710
XM_017010072.1
XP_016865561.1
3
670
XM_017010073.1
XP_016865562.1
3
461
XM_017010074.1
XP_016865563.1
3
727
XM_017010075.1
XP_016865564.1
3
386
XM_017010076.1
XP_016865565.1
3
372
XM_017010077.1
XP_016865566.1
2
228
GenBank mRNA
AB022663.1
-
3
354
AF060544.1
-
2
157
AK023698.1
-
2
156
AK023884.1
-
3
322
AK057868.1
-
3
356
AK129515.1
-
3
509
AK294262.1
-
3
325
AK295965.1
-
3
375
AW162926.1
-
3
409
BC126185.1
-
3
325
8991901
BC144061.1
-
3
325
9052581
BX647955.1
-
3
353
CB997918.1
-
2
214
CD106708.1
-
3
392
CR456702.1
-
1
118
DB456366.2
-
3
423
DC358984.1
-
3
386
DC396586.1
-
3
366
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
signal transduction
protein ubiquitination
androgen receptor signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of transcription, DNA-templated
regulation of androgen receptor signaling pathway
transcription coactivator activity
protein binding
zinc ion binding
ligase activity
ubiquitin-like protein transferase activity
ubiquitin conjugating enzyme binding
androgen receptor binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline