Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004291.3 | NP_004282.1 | 3 | 605 | ||
GenBank mRNA | BC029882.1 | - | 3 | 601 | 5171665 | |
U16826.1 | - | 3 | 493 |