Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 8 | 1234 | ||
NM_001286208.1 | NP_001273137.1 | 7 | 1179 | |||
NM_001286209.1 | NP_001273138.1 | 8 | 1126 | |||
NM_001481.2 | NP_001472.1 | 8 | 1111 | |||
XM_005256304.4 | XP_005256361.1 | 9 | 1766 | |||
XM_005256309.4 | XP_005256366.1 | 10 | 1683 | |||
XM_006721175.3 | XP_006721238.1 | 8 | 1164 | |||
XM_011522990.2 | XP_011521292.1 | 8 | 1185 | |||
XM_011522991.1 | XP_011521293.1 | 9 | 1340 | |||
XM_011522992.2 | XP_011521294.1 | 7 | 1133 | |||
XM_017023122.1 | XP_016878611.1 | 10 | 1491 | |||
XM_017023123.1 | XP_016878612.1 | 8 | 1190 | |||
XM_017023124.1 | XP_016878613.1 | 7 | 977 | |||
XM_017023125.1 | XP_016878614.1 | 7 | 1109 | |||
GenBank mRNA | AF050079.1 | - | 8 | 1110 | ||
AK293958.1 | - | 8 | 1141 | |||
AK294050.1 | - | 7 | 960 | |||
AK297857.1 | - | 8 | 1094 | |||
AK315261.1 | - | 8 | 1059 | |||
BC104785.1 | - | 8 | 1096 | 8143788 | ||
BC112121.1 | - | 8 | 1096 | 8327589 | ||
BX647947.1 | - | 7 | 1097 | |||
CR749499.1 | - | 6 | 1789 |