Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202469.1 | NP_001189398.1 | 8 | 881 | ||
NM_001483.2 | NP_001474.1 | 10 | 998 | |||
GenBank mRNA | AF029786.1 | - | 10 | 973 | ||
AJ001259.1 | - | 10 | 965 | |||
AK097049.1 | - | 8 | 867 | |||
BC000732.2 | - | 10 | 971 | 2959662 | ||
BC001837.2 | - | 10 | 971 | 2959662 | ||
BC030821.1 | - | 10 | 980 | 4808962 |