Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 9 | 1520 | ||
NM_001007241.2 | NP_001007242.2 | 8 | 1079 | |||
NM_001007242.2 | NP_001007243.2 | 7 | 1070 | |||
NM_001502.3 | NP_001493.2 | 8 | 1511 | |||
XM_017023155.1 | XP_016878644.1 | 8 | 1264 | |||
GenBank mRNA | AB035541.1 | - | 7 | 1300 | ||
AB035542.1 | - | 6 | 859 | |||
AK293384.1 | - | 8 | 1267 | |||
AK312277.1 | - | 8 | 1398 | |||
BC032693.2 | - | 6 | 917 | 5229838 | ||
BX647831.1 | - | 9 | 1408 | |||
BX648023.1 | - | 8 | 1405 | |||
BX648201.1 | - | 7 | 957 | |||
BX649067.1 | - | 8 | 966 | |||
CR749600.1 | - | 8 | 1470 | |||
U36221.1 | - | 8 | 1380 |