Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005335.1 | NP_001005335.1 | 3, 4 | 408 | ||
NM_001533.2 | NP_001524.2 | 3, 4 | 642 | |||
XM_011526886.1 | XP_011525188.1 | 3, 4 | 431 | |||
XM_011526887.1 | XP_011525189.1 | 3, 4 | 475 | |||
XM_011526889.1 | XP_011525191.1 | 3, 4 | 526 | |||
GenBank mRNA | AB044547.1 | - | 3, 4 | 637 | ||
AK292115.1 | - | 3, 4 | 407 | |||
AK298384.1 | - | 3, 4 | 478 | |||
AK301062.1 | - | 3, 4 | 416 | |||
AK313652.1 | - | 3, 4 | 577 | |||
BC069184.1 | - | 3, 4 | 581 | |||
BI256815.1 | - | 3, 4 | 408 | |||
X16135.1 | - | 3, 4 | 566 |