Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303265.1 | NP_001290194.1 | 6 | 773 | ||
NM_001545.2 | NP_001536.1 | 6 | 778 | |||
XM_017024574.1 | XP_016880063.1 | 6 | 771 | |||
GenBank mRNA | AK222553.1 | - | 6 | 768 | ||
BC015335.1 | - | 6 | 761 | 4418983 | ||
X81788.1 | - | 6 | 757 |