Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 11 | 1596 | ||
NM_001197079.1 | NP_001184008.1 | 10 | 1486 | |||
NM_001197080.1 | NP_001184009.1 | 10 | 1321 | |||
NM_001550.3 | NP_001541.2 | 10 | 1387 | |||
NR_120333.1 | 9 | 1149 | ||||
XM_011516142.2 | XP_011514444.1 | 11 | 1397 | |||
XM_011516143.2 | XP_011514445.1 | 10 | 1582 | |||
XM_017012151.1 | XP_016867640.1 | 10 | 1634 | |||
GenBank mRNA | AK297509.1 | - | 9 | 974 | ||
AK298894.1 | - | 10 | 1127 | |||
AK308066.1 | - | 11 | 1571 | |||
AK313211.1 | - | 11 | 1596 | |||
AK315967.1 | - | 10 | 1319 | |||
BC001272.1 | - | 10 | 1340 | 3458309 | ||
BT019889.1 | - | 10 | 1126 | |||
BT019890.1 | - | 10 | 1126 | |||
BX648799.1 | - | 2 | 824 |