Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001568.2 | NP_001559.1 | 6 | 600 | ||
XM_017013398.1 | XP_016868887.1 | 6 | 672 | |||
GenBank mRNA | AK124178.1 | - | 6 | 1204 | ||
AK313186.1 | - | 6 | 589 | |||
BC000734.2 | - | 6 | 576 | 2959855 | ||
BC005944.1 | - | 6 | 589 | |||
BC008419.1 | - | 6 | 594 | 4080571 | ||
BC016706.1 | - | 6 | 591 | 4096434 | ||
BC017887.1 | - | 6 | 596 | 4685684 | ||
BC021679.1 | - | 6 | 591 | 4132737 | ||
BG498300.1 | - | 6 | 603 | |||
CR457029.1 | - | 6 | 572 | |||
CR542275.1 | - | 6 | 572 | |||
U54562.1 | - | 6 | 578 | |||
U62962.1 | - | 6 | 594 | |||
U85947.1 | - | 6 | 580 | |||
U94175.1 | - | 6 | 592 |