Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025242.1 | NP_001020413.1 | 14 | 3313 | ||
NM_001025243.1 | NP_001020414.1 | 13 | 3166 | |||
NM_001569.3 | NP_001560.2 | 14 | 3403 | |||
XM_005274668.3 | XP_005274725.1 | 13 | 3335 | |||
XM_011531158.2 | XP_011529460.1 | 13 | 3091 | |||
GenBank mRNA | BC014963.1 | - | 13 | 3151 | 4906853 | |
BC054000.1 | - | 14 | 3345 | 6164719 | ||
L76191.1 | - | 14 | 3403 |