Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 3 | 2450 |
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002009.3 | NP_002000.1 | 4 | 2606 | ||
GenBank mRNA | M60828.1 | - | 4 | 2523 |