Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185075.1 | NP_001172004.1 | 16 | 3505 | ||
NM_001185076.1 | NP_001172005.1 | 16 | 3713 | |||
NM_001185081.1 | NP_001172010.1 | 15 | 3442 | |||
NM_001185082.1 | NP_001172011.1 | 16 | 3638 | |||
NM_002024.5 | NP_002015.1 | 17 | 3776 | |||
NR_033699.1 | 16 | 3580 | ||||
NR_033700.1 | 15 | 3517 | ||||
GenBank mRNA | AB209188.1 | - | 12 | 4201 | ||
BC086957.1 | - | 16 | 3669 | 30347992 | ||
M67468.1 | - | 16 | 3621 | |||
S65791.1 | - | 17 | 3765 | |||
X69962.1 | - | 17 | 3765 |