Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002031.2 | NP_002022.1 | 8 | 2491 | ||
XM_005266880.4 | XP_005266937.1 | 9 | 2563 | |||
XM_005266881.2 | XP_005266938.1 | 9 | 2480 | |||
XM_005266882.4 | XP_005266939.1 | 10 | 2792 | |||
XM_011535653.2 | XP_011533955.1 | 10 | 2795 | |||
XM_011535654.2 | XP_011533956.1 | 9 | 2577 | |||
XM_011535655.2 | XP_011533957.1 | 9 | 2574 | |||
XM_011535656.2 | XP_011533958.1 | 8 | 1948 | |||
XM_017010645.1 | XP_016866134.1 | 9 | 2681 | |||
GenBank mRNA | U00803.1 | - | 8 | 2491 | ||
U22322.1 | - | 8 | 2455 |