Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002048.2 | NP_002039.2 | 1 | 603 | ||
GenBank mRNA | BC074809.2 | - | 1 | 235 | 30915530 | |
BC074908.2 | - | 1 | 235 | 30915255 | ||
BC132682.1 | - | 1 | 278 | 40146704 | ||
L13698.1 | - | 1 | 603 |