Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282538.1 | NP_001269467.1 | 10 | 1360 | ||
NM_001282539.1 | NP_001269468.1 | 11 | 1457 | |||
NM_002074.4 | NP_002065.1 | 12 | 1506 | |||
XM_017001059.1 | XP_016856548.1 | 11 | 1410 | |||
XM_017001060.1 | XP_016856549.1 | 12 | 1276 | |||
XM_017001061.1 | XP_016856550.1 | 11 | 1449 | |||
GenBank mRNA | AF070597.1 | - | 4 | 475 | ||
AF070603.1 | - | 6 | 714 | |||
AK122949.1 | - | 10 | 1144 | |||
BC004186.2 | - | 12 | 1389 | 2964393 | ||
BC005888.2 | - | 10 | 1157 | 3605228 | ||
BC008991.1 | - | 12 | 1445 | 4155946 |