Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297571.1 | NP_001284500.1 | 10 | 1471 | ||
NM_002075.3 | NP_002066.1 | 10 | 1474 | |||
XM_011520953.2 | XP_011519255.1 | 9 | 2395 | |||
GenBank mRNA | AB209283.1 | - | 9 | 2615 | ||
AB593098.1 | - | 10 | 1400 | |||
AB593099.1 | - | 10 | 1399 | |||
BC000115.1 | - | 10 | 1387 | 3351520 | ||
BC002454.2 | - | 10 | 1394 | 3342882 | ||
BC015920.1 | - | 10 | 1224 | 4648399 | ||
M31328.1 | - | 9 | 1270 |