Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002087.3 | NP_002078.1 | 5 | 677 | ||
XM_005257253.1 | XP_005257310.1 | 5 | 722 | |||
GenBank mRNA | AF055008.1 | - | 5 | 483 | ||
AK000607.1 | - | 5 | 461 | |||
AK023348.1 | - | 3 | 225 | |||
AK222522.1 | - | 5 | 461 | |||
AK296090.1 | - | 5 | 415 | |||
AK303830.1 | - | 5 | 458 | |||
AY124489.1 | - | 4 | 439 | |||
BC000324.2 | - | 5 | 640 | 2821810 | ||
BC010577.2 | - | 4 | 431 | 3457813 | ||
BT006844.1 | - | 4 | 421 | |||
M75161.1 | - | 4 | 433 | |||
X62320.1 | - | 5 | 461 |