Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258040.2 | NP_001244969.1 | 4 | 773 | ||
NM_001289094.1 | NP_001276023.1 | 5 | 806 | |||
NM_002109.5 | NP_002100.2 | 5 | 893 | |||
GenBank mRNA | AK000498.1 | - | 5 | 573 | ||
AK055917.1 | - | 4 | 387 | |||
AK124831.1 | - | 4 | 2083 | |||
AK302295.1 | - | 4 | 387 | |||
AK307669.1 | - | 5 | 486 | |||
AY995220.1 | - | 5 | 470 | |||
BC011807.2 | - | 5 | 500 | 4331277 | ||
BC080514.1 | - | 5 | 498 | 2821065 | ||
X05345.1 | - | 5 | 547 | |||
Z11518.1 | - | 5 | 1028 |