Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318186.1 | NP_001305115.1 | 10 | 860 | ||
NM_001318187.1 | NP_001305116.1 | 10 | 860 | |||
NM_001318188.1 | NP_001305117.1 | 11 | 932 | |||
NM_002140.4 | NP_002131.2 | 11 | 1093 | |||
NM_031262.3 | NP_112552.1 | 11 | 1093 | |||
NM_031263.3 | NP_112553.1 | 11 | 932 | |||
XM_005251960.2 | XP_005252017.1 | 11 | 911 | |||
XM_005251963.3 | XP_005252020.1 | 10 | 873 | |||
XM_005251965.2 | XP_005252022.1 | 10 | 831 | |||
XM_011518616.1 | XP_011516918.1 | 10 | 873 | |||
XM_017014668.1 | XP_016870157.1 | 11 | 911 | |||
XM_017014669.1 | XP_016870158.1 | 10 | 831 | |||
GenBank mRNA | AB209562.1 | - | 10 | 789 | ||
AB451263.1 | - | 9 | 706 | |||
AB451390.1 | - | 9 | 706 | |||
AK096385.1 | - | 10 | 718 | |||
AK123117.1 | - | 5 | 1658 | |||
AK291336.1 | - | 11 | 915 | |||
AK294067.1 | - | 10 | 890 | |||
AK300744.1 | - | 10 | 803 | |||
AK307962.1 | - | 11 | 823 | |||
AY911506.1 | - | 10 | 769 | |||
BC000355.2 | - | 10 | 828 | 2964383 | ||
BC014980.1 | - | 11 | 899 | 4906241 | ||
BC025321.1 | - | 11 | 851 | |||
CR456771.1 | - | 9 | 706 | |||
S74678.1 | - | 10 | 813 | |||
X72727.1 | - | 11 | 915 |