Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258357.1 | NP_001245286.1 | 5 | 1222 | ||
NM_001258358.1 | NP_001245287.1 | 5 | 1234 | |||
NM_001258359.1 | NP_001245288.1 | 5 | 1628 | |||
NM_002149.3 | NP_002140.2 | 5 | 1448 | |||
NM_134421.2 | NP_602293.1 | 6 | 1590 | |||
XM_005246160.1 | XP_005246217.1 | 6 | 1513 | |||
XM_005246161.1 | XP_005246218.1 | 6 | 1636 | |||
XM_005246162.1 | XP_005246219.1 | 6 | 1639 | |||
XM_005246163.1 | XP_005246220.1 | 7 | 1686 | |||
XM_011510346.2 | XP_011508648.1 | 5 | 1292 | |||
XM_011510347.1 | XP_011508649.1 | 7 | 1733 | |||
XM_011510348.1 | XP_011508650.1 | 6 | 1526 | |||
XM_011510349.1 | XP_011508651.1 | 6 | 1552 | |||
XM_017003950.1 | XP_016859439.1 | 7 | 3180 | |||
XM_017003951.1 | XP_016859440.1 | 7 | 1663 | |||
XM_017003952.1 | XP_016859441.1 | 5 | 1408 | |||
XM_017003953.1 | XP_016859442.1 | 6 | 1543 | |||
GenBank mRNA | AF070616.1 | - | 3 | 817 | ||
AK000596.1 | - | 6 | 1596 | |||
AK093973.1 | - | 1 | 1953 | |||
BC009846.2 | - | 5 | 1423 | 4100091 | ||
BC017028.1 | - | 5 | 1415 | 3902811 | ||
BC017482.1 | - | 5 | 1438 | 3504942 | ||
CR749837.1 | - | 8 | 4732 |