Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286822.1 | NP_001273751.1 | 3 | 664 | ||
NM_004339.3 | NP_004330.1 | 6 | 992 | |||
NR_104597.1 | 5 | 883 | ||||
GenBank mRNA | AF149785.1 | - | 6 | 930 | ||
AK094882.1 | - | 7 | 697 | |||
AK095586.1 | - | 1 | 1292 | |||
AK290139.1 | - | 6 | 742 | |||
AK293318.1 | - | 3 | 393 | |||
AK298557.1 | - | 3 | 452 | |||
AK299204.1 | - | 4 | 551 | |||
BC000415.2 | - | 6 | 749 | 2961420 | ||
BC012858.1 | - | 6 | 775 | 3857505 | ||
BC019295.1 | - | 6 | 739 | 2823814 | ||
BC020983.1 | - | 6 | 756 | 3851447 | ||
BC031097.1 | - | 6 | 770 | 5288388 | ||
BC034250.1 | - | 6 | 764 | 3862830 | ||
L48984.1 | - | 3 | 390 | |||
Z50022.1 | - | 6 | 796 |