Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 3, 4 | 567 | ||
NM_199440.1 | NP_955472.1 | 3, 4 | 547 | |||
GenBank mRNA | AK297305.1 | - | 494 | |||
AK312240.1 | - | 3, 4 | 500 | |||
BC002676.2 | - | 3, 4 | 477 | 3609039 | ||
BC003030.1 | - | 3, 4 | 476 | 2821157 | ||
BC067082.1 | - | 3, 4 | 475 | 4025725 | ||
BC073746.1 | - | 3, 4 | 475 | 6372068 | ||
BG701476.1 | - | 3, 4 | 568 | |||
EU719626.1 | - | 2, 3 | 430 | |||
KU178139.1 | - | 2, 3 | 430 | |||
M22382.1 | - | 3, 4 | 475 | |||
M34664.1 | - | 3, 4 | 454 |