Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002186.2 | NP_002177.2 | 6 | 832 | ||
NM_176786.1 | NP_789743.2 | 7 | 921 | |||
XM_011531151.2 | XP_011529453.1 | 7 | 882 | |||
XM_011531152.2 | XP_011529454.1 | 7 | 858 | |||
XM_011531154.2 | XP_011529456.1 | 7 | 904 | |||
XM_011531155.2 | XP_011529457.1 | 7 | 891 | |||
XM_011531156.2 | XP_011529458.1 | 7 | 889 | |||
XM_011531157.2 | XP_011529459.1 | 7 | 886 | |||
XM_017029495.1 | XP_016884984.1 | 7 | 881 | |||
XM_017029497.1 | XP_016884986.1 | 7 | 928 | |||
XM_017029498.1 | XP_016884987.1 | 8 | 1033 | |||
XM_017029499.1 | XP_016884988.1 | 8 | 955 | |||
XM_017029500.1 | XP_016884989.1 | 7 | 895 | |||
XM_017029502.1 | XP_016884991.1 | 3 | 335 | |||
XM_017029503.1 | XP_016884992.1 | 8 | 934 | |||
GenBank mRNA | BC051337.1 | - | 7 | 796 | 6304829 | |
M84747.1 | - | 6 | 832 |