Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144996.1 | NP_001138468.1 | 2, 3 | 555 | ||
NM_002206.2 | NP_002197.2 | 2, 3 | 555 | |||
XM_005268839.1 | XP_005268896.1 | 2, 3 | 688 | |||
XM_005268840.1 | XP_005268897.1 | 2, 3 | 688 | |||
XM_005268841.1 | XP_005268898.1 | 2, 3 | 688 | |||
XM_005268842.1 | XP_005268899.1 | 2, 3 | 688 | |||
XM_005268844.1 | XP_005268901.1 | 3, 4 | 313 | |||
XM_005268846.1 | XP_005268903.1 | 3, 4 | 226 | |||
GenBank mRNA | AB210036.1 | - | 2, 3 | 525 | ||
AF032108.1 | - | 2, 3 | 496 | |||
AF052050.1 | - | 2, 3 | 335 | |||
AF072132.1 | - | 2, 3 | 335 | |||
AY358882.1 | - | 2, 3 | 354 | |||
BC050280.1 | - | 2, 3 | 551 | 5284443 |