Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002208.4 | NP_002199.3 | 21 | 2665 | ||
XM_011523823.2 | XP_011522125.1 | 21 | 3103 | |||
XM_011523825.2 | XP_011522127.1 | 21 | 3101 | |||
XM_011523827.2 | XP_011522129.1 | 21 | 3095 | |||
XM_011523828.2 | XP_011522130.1 | 21 | 3099 | |||
XM_017024586.1 | XP_016880075.1 | 22 | 4488 | |||
XM_017024587.1 | XP_016880076.1 | 21 | 4414 | |||
GenBank mRNA | BC113436.1 | - | 21 | 2655 | 8322488 | |
BC117207.1 | - | 21 | 2655 | 40125758 | ||
L25851.2 | - | 21 | 2691 |