Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114380.1 | NP_001107852.1 | 3 | 381 | ||
NM_002209.2 | NP_002200.2 | 3 | 381 | |||
XM_005255313.1 | XP_005255370.1 | 3 | 301 | |||
XM_006721044.1 | XP_006721107.1 | 3 | 332 | |||
XM_011545849.1 | XP_011544151.1 | 3 | 314 | |||
GenBank mRNA | AK298674.1 | - | 3 | 298 | ||
AK314246.1 | - | 3 | 335 | |||
BC008777.2 | - | 3 | 314 | 3142951 | ||
DC324373.1 | - | 3 | 381 | |||
Y00796.1 | - | 3 | 293 |