Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002214.2 | NP_002205.1 | 5 | 1431 | ||
XM_011515393.2 | XP_011513695.1 | 6 | 1234 | |||
XM_011515394.2 | XP_011513696.1 | 6 | 832 | |||
XM_011515396.1 | XP_011513698.1 | 6 | 1503 | |||
XM_017012178.1 | XP_016867667.1 | 6 | 1128 | |||
XM_017012179.1 | XP_016867668.1 | 6 | 1587 | |||
XM_017012180.1 | XP_016867669.1 | 6 | 1880 | |||
XM_017012181.1 | XP_016867670.1 | 7 | 1699 | |||
XM_017012182.1 | XP_016867671.1 | 6 | 814 | |||
XM_017012183.1 | XP_016867672.1 | 6 | 771 | |||
GenBank mRNA | AB209429.1 | - | 2 | 4250 | ||
AK295044.1 | - | 6 | 1503 | |||
BC002630.2 | - | 5 | 1431 | 3609490 | ||
BT006739.1 | - | 5 | 726 | |||
M73780.1 | - | 5 | 1406 |