Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002214.2 | NP_002205.1 | 8 | 1829 | ||
XM_011515393.2 | XP_011513695.1 | 9 | 1632 | |||
XM_011515394.2 | XP_011513696.1 | 9 | 1230 | |||
XM_011515396.1 | XP_011513698.1 | 9 | 1901 | |||
XM_017012178.1 | XP_016867667.1 | 9 | 1526 | |||
XM_017012179.1 | XP_016867668.1 | 9 | 1985 | |||
XM_017012180.1 | XP_016867669.1 | 9 | 2278 | |||
XM_017012181.1 | XP_016867670.1 | 10 | 2097 | |||
XM_017012182.1 | XP_016867671.1 | 9 | 1212 | |||
XM_017012183.1 | XP_016867672.1 | 9 | 1169 | |||
GenBank mRNA | AK295044.1 | - | 9 | 1901 | ||
BC002630.2 | - | 8 | 1829 | 3609490 | ||
BT006739.1 | - | 8 | 1124 | |||
M73780.1 | - | 8 | 1804 |