Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166434.2 | NP_001159906.1 | 2 | 187 | ||
NM_002215.3 | NP_002206.2 | 4 | 348 | |||
GenBank mRNA | AK292750.1 | - | 4 | 318 | ||
AK298398.1 | - | 4 | 347 | |||
AK298423.1 | - | 4 | 340 | |||
AK314198.1 | - | 4 | 318 | |||
BC069464.1 | - | 4 | 342 | 7262464 | ||
BX496392.1 | - | 4 | 349 | |||
DB188130.1 | - | 1 | 462 | |||
X16260.1 | - | 2 | 97 | |||
X63652.1 | - | 4 | 342 |