Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002216.2 | NP_002207.2 | 11 | 1341 | ||
GenBank mRNA | AK225277.1 | - | 13 | 1204 | ||
AK225329.1 | - | 11 | 1246 | |||
AK291379.1 | - | 11 | 1289 | |||
AK297334.1 | - | 6 | 532 | |||
BC132685.1 | - | 11 | 1228 | 40146707 | ||
CB154170.1 | - | 2 | 114 | |||
M18193.1 | - | 1 | 32 | |||
M33033.1 | - | 1 | 30 | |||
X07173.1 | - | 11 | 1242 |