Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 6 | 683 | ||
NM_001321852.1 | NP_001308781.1 | 5 | 686 | |||
NM_001321853.1 | NP_001308782.1 | 7 | 945 | |||
NM_001321854.1 | NP_001308783.1 | 6 | 861 | |||
NM_001321855.1 | NP_001308784.1 | 6 | 864 | |||
NM_001321856.1 | NP_001308785.1 | 5 | 599 | |||
NM_001321857.1 | NP_001308786.1 | 5 | 780 | |||
NM_002227.3 | NP_002218.2 | 5 | 780 | |||
XM_017001212.1 | XP_016856701.1 | 5 | 627 | |||
GenBank mRNA | AB209057.1 | - | 4 | 536 | ||
AB219242.1 | - | 3 | 430 | |||
AK314257.1 | - | 6 | 685 | |||
BC009079.1 | - | 5 | 715 | |||
BC023604.2 | - | 3 | 471 | |||
BC062431.1 | - | 5 | 715 | |||
BC111401.1 | - | 5 | 745 | |||
BC132729.1 | - | 4 | 498 | 40146751 | ||
BX648044.1 | - | 1 | 4101 | |||
HQ628631.1 | - | 3 | 466 | |||
M64174.1 | - | 4 | 505 |