Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 8 | 879 | ||
NM_001321852.1 | NP_001308781.1 | 7 | 882 | |||
NM_001321853.1 | NP_001308782.1 | 9 | 1141 | |||
NM_001321854.1 | NP_001308783.1 | 8 | 1057 | |||
NM_001321855.1 | NP_001308784.1 | 8 | 1060 | |||
NM_001321856.1 | NP_001308785.1 | 7 | 795 | |||
NM_001321857.1 | NP_001308786.1 | 7 | 976 | |||
NM_002227.3 | NP_002218.2 | 7 | 976 | |||
XM_017001212.1 | XP_016856701.1 | 7 | 823 | |||
GenBank mRNA | AB209057.1 | - | 6 | 732 | ||
AB219242.1 | - | 5 | 626 | |||
AK314257.1 | - | 8 | 881 | |||
BC009079.1 | - | 7 | 911 | |||
BC023604.2 | - | 5 | 667 | |||
BC062431.1 | - | 7 | 911 | |||
BC111401.1 | - | 7 | 941 | |||
BC132729.1 | - | 6 | 694 | 40146751 | ||
HQ628631.1 | - | 5 | 656 | |||
M64174.1 | - | 6 | 701 |