Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024844.1 | NP_001020015.1 | 9 | 946 | ||
NM_002231.3 | NP_002222.1 | 10 | 1021 | |||
XM_006718223.2 | XP_006718286.1 | 11 | 1060 | |||
XM_011520067.2 | XP_011518369.1 | 7 | 723 | |||
GenBank mRNA | AK225410.1 | - | 10 | 947 | ||
AK225708.1 | - | 10 | 975 | |||
AK312677.1 | - | 10 | 947 | |||
AY303776.1 | - | 7 | 698 | |||
BC000726.2 | - | 10 | 919 | 2959683 | ||
BC001821.2 | - | 10 | 919 | 2959683 | ||
BT019761.1 | - | 8 | 773 | |||
BT019762.1 | - | 8 | 773 | |||
CR407650.1 | - | 8 | 773 | |||
CR542255.1 | - | 8 | 773 | |||
U20770.1 | - | 10 | 938 |